Product Details

SNP ID
rs9131
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:74097332 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATTTCCCTGCCGTCACATTGATCT[C/T]ACTGGCCATTTTCTTGGATTCCTCA
Phenotype
MIM: 139110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL2 PubMed Links

Gene Details

Gene
CXCL2
Gene Name
C-X-C motif chemokine ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002089.3 Intron NP_002080.1

View Full Product Details