Product Details

SNP ID
rs1497568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:46038741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTACGCCATTCTTTCTGATCACAC[A/G]TTTAAATAAAACTGATCAAAATACC
Phenotype
MIM: 137166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRG1 PubMed Links

Gene Details

Gene
GABRG1
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173536.3 3797 UTR 3 NP_775807.2
XM_017007990.1 3797 UTR 3 XP_016863479.1

View Full Product Details