Product Details

SNP ID
rs907522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7053530 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGGCCCTTGCTTTTCCAGCGGG[C/T]GGCAACATGGTAGAACTGGGTCCCC
Phenotype
MIM: 606173 MIM: 608790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRPEL1 PubMed Links

Gene Details

Gene
GRPEL1
Gene Name
GrpE like 1, mitochondrial
There are no transcripts associated with this gene.

Gene
LOC100129931
Gene Name
uncharacterized LOC100129931
There are no transcripts associated with this gene.

Gene
TADA2B
Gene Name
transcriptional adaptor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152293.2 Intron NP_689506.2
XM_011513595.1 Intron XP_011511897.1

View Full Product Details