Product Details

SNP ID
rs1062682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24532993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATAGCTGCTGGCGTACATTGTC[C/T]GCGGACATCAGGGACCTGTAGTTAA
Phenotype
MIM: 603403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX15 PubMed Links

Gene Details

Gene
DHX15
Gene Name
DEAH-box helicase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001358.2 2139 Silent Mutation GCA,GCG A657A NP_001349.2

View Full Product Details