Product Details

SNP ID
rs1351382
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32854519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACGCGAGGCTGGTGGAAAAAG[C/G]GGGTGCTTTGACTCTTAGCTGGAAG
Phenotype
MIM: 177046 MIM: 177045 MIM: 170260
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PSMB8 PubMed Links
Additional Information
For this assay, SNP(s) [rs111240027] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PSMB8
Gene Name
proteasome subunit beta 8
There are no transcripts associated with this gene.

Gene
PSMB8-AS1
Gene Name
PSMB8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PSMB9
Gene Name
proteasome subunit beta 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002800.4 Intron NP_002791.1
Gene
TAP1
Gene Name
transporter 1, ATP binding cassette subfamily B member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000593.5 Intron NP_000584.2
NM_001292022.1 Intron NP_001278951.1

View Full Product Details