Product Details

SNP ID
rs1485522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176076787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATTCCTACTCCTGAATGAGAGG[C/T]GGCCCATATAGGCTATTGATTACCA
Phenotype
MIM: 609005
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR17 PubMed Links

Gene Details

Gene
WDR17
Gene Name
WD repeat domain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170710.4 Intron NP_733828.2
NM_181265.3 Intron NP_851782.3
XM_005262725.3 Intron XP_005262782.1
XM_011531582.2 Intron XP_011529884.1
XM_017007686.1 Intron XP_016863175.1
XM_017007687.1 Intron XP_016863176.1
XM_017007688.1 Intron XP_016863177.1
XM_017007689.1 Intron XP_016863178.1
XM_017007690.1 Intron XP_016863179.1
XM_017007691.1 Intron XP_016863180.1
XM_017007692.1 Intron XP_016863181.1
XM_017007693.1 Intron XP_016863182.1
XM_017007694.1 Intron XP_016863183.1
XM_017007695.1 Intron XP_016863184.1
XM_017007696.1 Intron XP_016863185.1
XM_017007697.1 Intron XP_016863186.1

View Full Product Details