Product Details

SNP ID
rs1802545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61485899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTTCTGGTTCTCTAGCAGCTG[A/G]GACATTTCTCTGATAATCAATGAGA
Phenotype
MIM: 602559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPO1 PubMed Links

Gene Details

Gene
XPO1
Gene Name
exportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003400.3 3105 Missense Mutation CCA,TCA P793S NP_003391.1
XM_005264544.1 3105 Missense Mutation CCA,TCA P778S XP_005264601.1
XM_005264546.1 3105 Missense Mutation CCA,TCA P662S XP_005264603.1
XM_006712094.2 3105 Missense Mutation CCA,TCA P793S XP_006712157.1
XM_011533097.1 3105 Missense Mutation CCA,TCA P793S XP_011531399.1
XM_011533098.1 3105 Missense Mutation CCA,TCA P748S XP_011531400.1
XM_011533099.2 3105 Missense Mutation CCA,TCA P727S XP_011531401.1

View Full Product Details