Product Details

SNP ID
rs1177227
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61028790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTTAGAAAAGAAAGGAAACAAA[A/C]CAACCAAATGCAGTGTGTGAGCCTT
Phenotype
MIM: 601789
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PEX13 PubMed Links

Gene Details

Gene
PEX13
Gene Name
peroxisomal biogenesis factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002618.3 Intron NP_002609.1

View Full Product Details