Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130480.2 | Intron | NP_001123952.1 | |||
NM_001130481.2 | Intron | NP_001123953.1 | |||
NM_001130482.2 | Intron | NP_001123954.1 | |||
NM_001130483.2 | Intron | NP_001123955.1 | |||
NM_001288836.1 | Intron | NP_001275765.1 | |||
NM_001288837.1 | Intron | NP_001275766.1 | |||
NM_001288838.1 | Intron | NP_001275767.1 | |||
NM_001288839.1 | Intron | NP_001275768.1 | |||
NM_006296.6 | Intron | NP_006287.2 | |||
XM_005264540.4 | Intron | XP_005264597.1 | |||
XM_006712090.3 | Intron | XP_006712153.1 | |||
XM_006712091.3 | Intron | XP_006712154.1 | |||
XM_006712092.3 | Intron | XP_006712155.1 | |||
XM_006712093.3 | Intron | XP_006712156.1 | |||
XM_011533092.2 | Intron | XP_011531394.1 | |||
XM_017004858.1 | Intron | XP_016860347.1 | |||
XM_017004859.1 | Intron | XP_016860348.1 | |||
XM_017004860.1 | Intron | XP_016860349.1 | |||
XM_017004861.1 | Intron | XP_016860350.1 | |||
XM_017004862.1 | Intron | XP_016860351.1 | |||
XM_017004863.1 | Intron | XP_016860352.1 | |||
XM_017004864.1 | Intron | XP_016860353.1 | |||
XM_017004865.1 | Intron | XP_016860354.1 | |||
XM_017004866.1 | Intron | XP_016860355.1 |