Product Details

SNP ID
rs814771
Assay Type
Functionally tested
NCBI dbSNP Submissions
58
Location
Chr.1:110339822 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCAAAAATTCTTCGGGCGGGGGCGA[A/G]TCGCGCAGCAGCTCCCGGGGTGGAG
Phenotype
MIM: 606077
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC440600 PubMed Links

Gene Details

Gene
LOC440600
Gene Name
uncharacterized LOC440600
There are no transcripts associated with this gene.

Gene
RBM15
Gene Name
RNA binding motif protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201545.1 500 Silent Mutation GAA,GAG E139E NP_001188474.1
NM_022768.4 500 Silent Mutation GAA,GAG E139E NP_073605.4

View Full Product Details