Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281765.2 | 4815 | UTR 3 | NP_001268694.1 | ||
NM_001281766.2 | 4815 | UTR 3 | NP_001268695.1 | ||
NM_001281767.2 | 4815 | Intron | NP_001268696.1 | ||
NM_001318761.1 | 4815 | UTR 3 | NP_001305690.1 | ||
NM_004439.7 | 4815 | UTR 3 | NP_004430.4 | ||
NM_182472.4 | 4815 | UTR 3 | NP_872272.2 | ||
XM_005265653.3 | 4815 | UTR 3 | XP_005265710.1 | ||
XM_011531735.2 | 4815 | UTR 3 | XP_011530037.1 | ||
XM_017007878.1 | 4815 | UTR 3 | XP_016863367.1 | ||
XM_017007879.1 | 4815 | Intron | XP_016863368.1 | ||
XM_017007880.1 | 4815 | UTR 3 | XP_016863369.1 | ||
XM_017007881.1 | 4815 | Intron | XP_016863370.1 |