Product Details

SNP ID
rs1048725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117710996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATAAAACTAGAATTTAACAAGC[A/G]AAATACTTAATATGGCTTTTAATGG
Phenotype
MIM: 609772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTTNBP2 PubMed Links

Gene Details

Gene
CTTNBP2
Gene Name
cortactin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033427.2 5625 UTR 3 NP_219499.1
XM_005250634.4 5625 UTR 3 XP_005250691.1
XM_005250635.3 5625 UTR 3 XP_005250692.1
XM_011516615.2 5625 UTR 3 XP_011514917.1
XM_011516616.2 5625 Intron XP_011514918.1
XM_011516618.2 5625 Intron XP_011514920.1
XM_017012706.1 5625 Intron XP_016868195.1
XM_017012707.1 5625 Intron XP_016868196.1
XM_017012708.1 5625 UTR 3 XP_016868197.1

View Full Product Details