Product Details

SNP ID
rs1149056
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:32224049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTCCAGATCGTCAATGTATGG[C/T]ACTCTGTGAATGGTGAGAGGCTGGG
Phenotype
MIM: 603911
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCDC2B PubMed Links

Gene Details

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003757.3 645 Missense Mutation CAC,TAC H38Y NP_003748.1
XM_017002671.1 645 UTR 5 XP_016858160.1
Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
There are no transcripts associated with this gene.

View Full Product Details