Product Details

SNP ID
rs859537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38978008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAGACAACATATGATGGATTGC[G/T]ATCTCTCAGCGCAGCAGGACACGGG
Phenotype
MIM: 609062
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POU6F2 PubMed Links

Gene Details

Gene
POU6F2
Gene Name
POU class 6 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166018.1 131 Intron NP_001159490.1
NM_007252.3 131 Intron NP_009183.3
XM_011515113.2 131 Missense Mutation GAT,TAT D19Y XP_011513415.1
Gene
POU6F2-AS2
Gene Name
POU6F2 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details