Product Details
- SNP ID
-
rs1229026
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:38984862 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTTAGAGAAACAGGTATATTTT[A/T]AAAAGAATACATACCTGTTAAATGG
- Phenotype
-
MIM: 609062
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
POU6F2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116707964] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POU6F2
- Gene Name
- POU class 6 homeobox 2
- Gene
- POU6F2-AS2
- Gene Name
- POU6F2 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details