Product Details
- SNP ID
-
rs1051162
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:39566508 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAGAAGCAGACGAGTCGCTCCTGGC[A/G]CAGCGGGAATGGCAGAGTAACATGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105375240
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7777434] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105375240
- Gene Name
- uncharacterized LOC105375240
There are no transcripts associated with this gene.
- Gene
- YAE1D1
- Gene Name
- Yae1 domain containing 1
View Full Product Details