Product Details

SNP ID
rs1038426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67737817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGAGAAACCAAGATAGAATCTC[A/C]AATATCTGATATATTAGATTGACTC
Phenotype
MIM: 138850
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GNRHR PubMed Links

Gene Details

Gene
GNRHR
Gene Name
gonadotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000406.2 5401 UTR 3 NP_000397.1
NM_001012763.1 5401 UTR 3 NP_001012781.1

View Full Product Details