Product Details
- SNP ID
-
rs1511822
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:23440594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACGGGGCATGGGCCGGGTACCC[A/G]GAGTTGCTGGCCATGGGGCAGATGC
- Phenotype
-
MIM: 612022
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTUD1
PubMed Links
Gene Details
- Gene
- OTUD1
- Gene Name
- OTU deubiquitinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145373.2 |
1326 |
Silent Mutation |
CCA,CCG |
P379P |
NP_001138845.1 |
View Full Product Details