Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000858.5 | Intron | NP_000849.1 | |||
NM_001159390.1 | Intron | NP_001152862.1 | |||
NM_001159391.1 | Intron | NP_001152863.1 | |||
NM_001242839.1 | Intron | NP_001229768.1 | |||
NM_001242840.1 | Intron | NP_001229769.1 | |||
XM_005273103.3 | Intron | XP_005273160.1 | |||
XM_005273104.4 | Intron | XP_005273161.1 | |||
XM_005273105.3 | Intron | XP_005273162.1 | |||
XM_005273106.2 | Intron | XP_005273163.1 | |||
XM_011544174.2 | Intron | XP_011542476.1 | |||
XM_017001091.1 | Intron | XP_016856580.1 | |||
XM_017001092.1 | Intron | XP_016856581.1 | |||
XM_017001093.1 | Intron | XP_016856582.1 | |||
XM_017001094.1 | Intron | XP_016856583.1 | |||
XM_017001095.1 | Intron | XP_016856584.1 | |||
XM_017001096.1 | Intron | XP_016856585.1 |