Product Details

SNP ID
rs1049452
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:203179133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGTCCTTTGATAAGGAGGGCT[A/G]GGGGGCAGGGAGTTGAAGAAATTCC
Phenotype
MIM: 601525 MIM: 160795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHI3L1 PubMed Links

Gene Details

Gene
CHI3L1
Gene Name
chitinase 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276.2 1635 UTR 3 NP_001267.2
Gene
MYBPH
Gene Name
myosin binding protein H
There are no transcripts associated with this gene.

View Full Product Details