Product Details
- SNP ID
-
rs1475263
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:83873814 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGATAGAAATTAACCTTGAAAACAT[A/T]AGCTACACCCACCTTCCTTTTCCTC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TTLL7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144171714] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTLL7
- Gene Name
- tubulin tyrosine ligase like 7
View Full Product Details