Product Details

SNP ID
rs1055362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101578976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTCCCAGCACCACCAGCTGCCC[A/G]GGAAGCAGGACGGATAGAGGGAACA
Phenotype
MIM: 616467 MIM: 606343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
POLL
Gene Name
polymerase (DNA) lambda
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174084.1 2700 UTR 3 NP_001167555.1
NM_001174085.1 2700 UTR 3 NP_001167556.1
NM_001308382.1 2700 UTR 3 NP_001295311.1
NM_013274.3 2700 UTR 3 NP_037406.1
XM_006717775.1 2700 UTR 3 XP_006717838.1
XM_006717776.1 2700 UTR 3 XP_006717839.1
XM_006717777.1 2700 UTR 3 XP_006717840.1
XM_011539650.1 2700 UTR 3 XP_011537952.1
XM_011539651.1 2700 UTR 3 XP_011537953.1
XM_011539652.1 2700 UTR 3 XP_011537954.1
XM_011539653.1 2700 UTR 3 XP_011537955.1
XM_011539654.1 2700 UTR 3 XP_011537956.1
XM_011539655.1 2700 UTR 3 XP_011537957.1
XM_011539656.1 2700 UTR 3 XP_011537958.1
XM_011539657.1 2700 UTR 3 XP_011537959.1
XM_011539659.1 2700 UTR 3 XP_011537961.1
XM_011539660.1 2700 UTR 3 XP_011537962.1
XM_011539662.1 2700 UTR 3 XP_011537964.1
XM_011539663.1 2700 UTR 3 XP_011537965.1
XM_011539664.1 2700 UTR 3 XP_011537966.1
XM_011539665.2 2700 UTR 3 XP_011537967.1
XM_011539666.1 2700 UTR 3 XP_011537968.1
XM_011539667.1 2700 UTR 3 XP_011537969.1
XM_017016084.1 2700 UTR 3 XP_016871573.1
XM_017016085.1 2700 UTR 3 XP_016871574.1
XM_017016086.1 2700 UTR 3 XP_016871575.1
XM_017016087.1 2700 UTR 3 XP_016871576.1
XM_017016088.1 2700 Intron XP_016871577.1
XM_017016089.1 2700 UTR 3 XP_016871578.1
XM_017016090.1 2700 UTR 3 XP_016871579.1
XM_017016091.1 2700 UTR 3 XP_016871580.1
XM_017016092.1 2700 Intron XP_016871581.1
XM_017016093.1 2700 UTR 3 XP_016871582.1

View Full Product Details