Product Details

SNP ID
rs1530589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101858276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATGGCATAGTGGTTAAGAGCA[C/T]GGCTCTAGAGTTAGCCTGCCTGGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf76 PubMed Links

Gene Details

Gene
C10orf76
Gene Name
chromosome 10 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024541.2 Intron NP_078817.2
XM_006717969.1 Intron XP_006718032.1
XM_011540151.2 Intron XP_011538453.1
XM_011540152.2 Intron XP_011538454.1
XM_011540153.2 Intron XP_011538455.1
XM_011540154.2 Intron XP_011538456.1
XM_011540156.2 Intron XP_011538458.1
XM_011540157.2 Intron XP_011538459.1
XM_017016635.1 Intron XP_016872124.1
XM_017016636.1 Intron XP_016872125.1

View Full Product Details