Product Details

SNP ID
rs932959
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:41518241 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTAGGAATGTATATACATACATAA[C/G]GTGTGTGGATAACGATCACATGAAA
Phenotype
MIM: 151520 MIM: 611475
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LTK PubMed Links

Gene Details

Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
RPAP1
Gene Name
RNA polymerase II associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015540.3 Intron NP_056355.2
XM_005254297.1 Intron XP_005254354.1
XM_006720469.1 Intron XP_006720532.1

View Full Product Details