Product Details

SNP ID
rs1023194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41563531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGTGCAGGTCCTGTTCAGGAGG[C/T]GCTGAGTTGGAGCAGATTAGAGAGG
Phenotype
MIM: 600341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TYRO3 PubMed Links

Gene Details

Gene
TYRO3
Gene Name
TYRO3 protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006293.3 Intron NP_006284.2
XM_017022542.1 Intron XP_016878031.1
XM_017022543.1 Intron XP_016878032.1

View Full Product Details