Product Details

SNP ID
rs1706024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40056443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAACAGCTCCCGCCTTCCCCCC[A/C]AGCAGAACCCGACACTCGAAGAAGG
Phenotype
MIM: 611963
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC344967 PubMed Links

Gene Details

Gene
LOC344967
Gene Name
acyl-CoA thioesterase 7 pseudogene
There are no transcripts associated with this gene.

Gene
N4BP2
Gene Name
NEDD4 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318359.1 221 Intron NP_001305288.1
NM_018177.5 221 Intron NP_060647.2
XM_006714023.3 221 Intron XP_006714086.1
XM_011513716.2 221 Missense Mutation CAA,CCA Q13P XP_011512018.1
XM_011513717.2 221 Missense Mutation CAA,CCA Q13P XP_011512019.1
XM_011513718.2 221 Intron XP_011512020.1
XM_011513719.2 221 Intron XP_011512021.1
XM_011513720.1 221 Intron XP_011512022.1
XM_011513721.1 221 Intron XP_011512023.1
XM_017008397.1 221 Missense Mutation CAA,CCA Q13P XP_016863886.1
XM_017008398.1 221 Intron XP_016863887.1
XM_017008399.1 221 Intron XP_016863888.1

View Full Product Details