Product Details

SNP ID
rs1452393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73875365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGGGAAGCTTAAGCAAAGTCAA[C/T]GACCTTCCCTGGGCTTTATTGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D21 PubMed Links

Gene Details

Gene
TBC1D21
Gene Name
TBC1 domain family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286434.1 Intron NP_001273363.1
NM_153356.2 Intron NP_699187.1
XM_006720409.3 Intron XP_006720472.1
XM_006720410.3 Intron XP_006720473.1
XM_006720411.3 Intron XP_006720474.1
XM_011521281.2 Intron XP_011519583.1
XM_011521282.2 Intron XP_011519584.1
XM_011521283.2 Intron XP_011519585.1
XM_011521284.2 Intron XP_011519586.1

View Full Product Details