Product Details
- SNP ID
-
rs1414918
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:96054718 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCTTAAATGGATTGGCCAACCTC[A/G]CTACCTTGGTTTTTGAGCAGTGAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNJ
PubMed Links
Gene Details
- Gene
- CCNJ
- Gene Name
- cyclin J
- Gene
- ENTPD1-AS1
- Gene Name
- ENTPD1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MIR3157
- Gene Name
- microRNA 3157
There are no transcripts associated with this gene.
View Full Product Details