Product Details

SNP ID
rs14674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102254012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTCAGAACATTTACATTTTTT[C/T]CCCCTCAGCAAAAAGATAAATCACA
Phenotype
MIM: 608413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR5 PubMed Links

Gene Details

Gene
UBR5
Gene Name
ubiquitin protein ligase E3 component n-recognin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282873.1 9118 UTR 3 NP_001269802.1
NM_015902.5 9118 UTR 3 NP_056986.2
XM_005250962.4 9118 UTR 3 XP_005251019.1
XM_011517104.2 9118 UTR 3 XP_011515406.2
XM_011517105.2 9118 UTR 3 XP_011515407.2
XM_011517106.2 9118 UTR 3 XP_011515408.2
XM_017013533.1 9118 UTR 3 XP_016869022.1
XM_017013534.1 9118 UTR 3 XP_016869023.1
Gene
UBR5-AS1
Gene Name
UBR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details