Product Details

SNP ID
rs1334892
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:97321224 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGGGAAAGCGCGGACGTGCTGGGCG[G/T]CTGTGCTTCGGTAGGCGACCACCGC
Phenotype
MIM: 602503
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FRAT1 PubMed Links

Gene Details

Gene
FRAT1
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005479.3 1960 UTR 3 NP_005470.2

View Full Product Details