Product Details
- SNP ID
-
rs1764305
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:96133148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACATTTTATTAGAAATCCAAATA[C/T]AACTTTAACTATCATCCTTCTGAAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF518A
PubMed Links
Gene Details
- Gene
- ZNF518A
- Gene Name
- zinc finger protein 518A
View Full Product Details