Product Details

SNP ID
rs1397573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71126157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTTTTGAAGGGAGGATGGAGGA[A/G]GGCAAAGTGACATTTATGCTGAGTC
Phenotype
MIM: 600769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN8 PubMed Links

Gene Details

Gene
TSPAN8
Gene Name
tetraspanin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004616.2 Intron NP_004607.1
XM_006719583.3 Intron XP_006719646.1
XM_017019913.1 Intron XP_016875402.1

View Full Product Details