Product Details

SNP ID
rs3144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77613862 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCAGGTCTGGGCTTCTGAGGTCC[C/T]GGTAGAAGGAGGGCAGGTGGTGAGG
Phenotype
MIM: 609791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINGO1 PubMed Links

Gene Details

Gene
LINGO1
Gene Name
leucine rich repeat and Ig domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301186.1 2239 UTR 3 NP_001288115.1
NM_001301187.1 2239 UTR 3 NP_001288116.1
NM_001301189.1 2239 UTR 3 NP_001288118.1
NM_001301191.1 2239 UTR 3 NP_001288120.1
NM_001301192.1 2239 UTR 3 NP_001288121.1
NM_001301194.1 2239 UTR 3 NP_001288123.1
NM_001301195.1 2239 UTR 3 NP_001288124.1
NM_001301197.1 2239 UTR 3 NP_001288126.1
NM_001301198.1 2239 UTR 3 NP_001288127.1
NM_001301199.1 2239 UTR 3 NP_001288128.1
NM_001301200.1 2239 UTR 3 NP_001288129.1
NM_032808.6 2239 UTR 3 NP_116197.4
XM_011522118.2 2239 UTR 3 XP_011520420.1
XM_017022682.1 2239 UTR 3 XP_016878171.1

View Full Product Details