Product Details

SNP ID
rs1386801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55020746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTAGGGCTATGCAAGGACGGAG[C/G]GGGAGGCAGGGAAAGGAAAACAGAA
Phenotype
MIM: 611635
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NEUROD4 PubMed Links

Gene Details

Gene
NEUROD4
Gene Name
neuronal differentiation 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021191.2 Intron NP_067014.2

View Full Product Details