Product Details
- SNP ID
-
rs881346
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:132186469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCTGCGAAGGAGCCGGCTTTGG[C/G]CAAGGTGCCTTCCTGGACGGGTGTG
- Phenotype
-
MIM: 608407
MIM: 611198
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DPYSL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138873687] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPYSL4
- Gene Name
- dihydropyrimidinase like 4
- Gene
- JAKMIP3
- Gene Name
- Janus kinase and microtubule interacting protein 3
There are no transcripts associated with this gene.
View Full Product Details