Product Details

SNP ID
rs13789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79360717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATTTCAAATTTTAAAATTGCT[C/G]CACCATAAGATGAATAAAGAGCTTA
Phenotype
MIM: 300222
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITM2A PubMed Links

Gene Details

Gene
ITM2A
Gene Name
integral membrane protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171581.1 1501 UTR 3 NP_001165052.1
NM_004867.4 1501 UTR 3 NP_004858.1

View Full Product Details