Product Details

SNP ID
rs1802053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73762462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGAAAAATTTAAGACTGGAAT[A/G]ACAGATTGTTTTTCCTGCAATGCTG
Phenotype
MIM: 603092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP11 PubMed Links

Gene Details

Gene
DUSP11
Gene Name
dual specificity phosphatase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003584.2 1375 UTR 3 NP_003575.2

View Full Product Details