Product Details

SNP ID
rs702614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53889019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATGCAACATCCAAAATCTAGTAC[A/G]TAAAGGAAGTTGTGTCCCTGATACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL15 PubMed Links

Gene Details

Gene
ARL15
Gene Name
ADP ribosylation factor like GTPase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019087.2 Intron NP_061960.1
XM_011543498.2 Intron XP_011541800.1
XM_011543499.2 Intron XP_011541801.1
XM_011543500.2 Intron XP_011541802.1
XM_017009598.1 Intron XP_016865087.1

View Full Product Details