Product Details
- SNP ID
-
rs913038
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:15106548 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCAAGTCTTCCATTATCTCCAAGA[A/G]AGGTCCTATATTATGTACTACTGTG
- Phenotype
-
MIM: 603801
MIM: 606116
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf111
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7901643] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C10orf111
- Gene Name
- chromosome 10 open reading frame 111
There are no transcripts associated with this gene.
- Gene
- NMT2
- Gene Name
- N-myristoyltransferase 2
- Gene
- RPP38
- Gene Name
- ribonuclease P/MRP subunit p38
There are no transcripts associated with this gene.
View Full Product Details