Product Details

SNP ID
rs1047689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55164231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGATCTATGAGAATGCCATTGC[A/G]TTTCTAATAGAACAGAGAAATGTCT
Phenotype
MIM: 613131 MIM: 613132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145734.2 456 Intron NP_001139206.2
NM_001170402.1 456 Intron NP_001163873.1
NM_152623.2 456 Intron NP_689836.2
XM_011543218.2 456 Intron XP_011541520.1
Gene
GPX8
Gene Name
glutathione peroxidase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008397.3 456 UTR 3 NP_001008398.2
NM_001306197.1 456 UTR 3 NP_001293126.1
NM_001306198.1 456 UTR 3 NP_001293127.1
NM_001306201.1 456 UTR 3 NP_001293130.1
XM_006714631.2 456 UTR 3 XP_006714694.1
Gene
MIR449A
Gene Name
microRNA 449a
There are no transcripts associated with this gene.

Gene
MIR449B
Gene Name
microRNA 449b
There are no transcripts associated with this gene.

Gene
MIR449C
Gene Name
microRNA 449c
There are no transcripts associated with this gene.

View Full Product Details