Product Details
- SNP ID
-
rs829257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:58969539 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTCAGATGCAAATCACTTTCTCAA[A/G]GCCTCAAGGAAGCCTTCTGTGGCCC
- Phenotype
-
MIM: 600129
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDE4D
PubMed Links
Gene Details
- Gene
- PDE4D
- Gene Name
- phosphodiesterase 4D
View Full Product Details