Product Details
- SNP ID
-
rs726216
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23603594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATGCAGCATCCACAGCTGTGGAC[C/T]TGAGGAAAGCTGATGCCCTGTCTTA
- Phenotype
-
MIM: 616543
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST9
PubMed Links
Gene Details
- Gene
- CST9
- Gene Name
- cystatin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008693.2 |
505 |
Silent Mutation |
CAA,CAG |
Q132Q |
NP_001008693.2 |
View Full Product Details