Product Details
- SNP ID
-
rs7492
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23079620 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAATACTTACATTCTCATTTAGC[A/G]TTCATGAAGTGATTACTGACATTTA
- Phenotype
-
MIM: 120577
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD93
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41492545] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD93
- Gene Name
- CD93 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012072.3 |
6437 |
UTR 3 |
|
|
NP_036204.2 |
View Full Product Details