Product Details
- SNP ID
-
rs8565
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:76000956 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGCGGCACAACTGCGGCTGATACC[C/T]TGGGTGGAAAAGATCAGAATGACAG
- Phenotype
-
MIM: 616695
MIM: 616550
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STYXL1
PubMed Links
Gene Details
- Gene
- STYXL1
- Gene Name
- serine/threonine/tyrosine interacting-like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317785.1 |
940 |
Silent Mutation |
CAA,CAG |
Q248Q |
NP_001304714.1 |
NM_001317786.1 |
940 |
Silent Mutation |
CAA,CAG |
Q248Q |
NP_001304715.1 |
NM_001317787.1 |
940 |
Silent Mutation |
CAA,CAG |
Q152Q |
NP_001304716.1 |
NM_001317788.1 |
940 |
Intron |
|
|
NP_001304717.1 |
NM_001317789.1 |
940 |
Silent Mutation |
CAA,CAG |
Q110Q |
NP_001304718.1 |
NM_016086.2 |
940 |
Silent Mutation |
CAA,CAG |
Q248Q |
NP_057170.1 |
XM_011516292.2 |
940 |
Intron |
|
|
XP_011514594.1 |
XM_011516293.2 |
940 |
Silent Mutation |
CAA,CAG |
Q248Q |
XP_011514595.1 |
XM_011516300.1 |
940 |
Silent Mutation |
CAA,CAG |
Q110Q |
XP_011514602.1 |
XM_017012296.1 |
940 |
Silent Mutation |
CAA,CAG |
Q286Q |
XP_016867785.1 |
XM_017012297.1 |
940 |
Intron |
|
|
XP_016867786.1 |
XM_017012298.1 |
940 |
Intron |
|
|
XP_016867787.1 |
XM_017012299.1 |
940 |
Intron |
|
|
XP_016867788.1 |
XM_017012300.1 |
940 |
Silent Mutation |
CAA,CAG |
Q170Q |
XP_016867789.1 |
XM_017012301.1 |
940 |
Silent Mutation |
CAA,CAG |
Q110Q |
XP_016867790.1 |
XM_017012302.1 |
940 |
Silent Mutation |
CAA,CAG |
Q110Q |
XP_016867791.1 |
- Gene
- TMEM120A
- Gene Name
- transmembrane protein 120A
There are no transcripts associated with this gene.
View Full Product Details