Product Details
- SNP ID
-
rs9909
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47778223 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAAGGCTATACAAGCAAGTAGTATA[C/G]AGATTGTTTCCTTAAAAGATAAATT
- Phenotype
-
MIM: 607614
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NUP160
PubMed Links
Gene Details
- Gene
- NUP160
- Gene Name
- nucleoporin 160
View Full Product Details