Product Details

SNP ID
rs1050754
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110293952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAACAAACTTTGAGGGAACCG[G/T]GGTACTGATAATGAGAAAAGTTAAA
Phenotype
MIM: 600020 MIM: 603519
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MXI1 PubMed Links

Gene Details

Gene
MXI1
Gene Name
MAX interactor 1, dimerization protein
There are no transcripts associated with this gene.

Gene
SMNDC1
Gene Name
survival motor neuron domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005871.3 1117 UTR 3 NP_005862.1
XM_005269382.3 1117 UTR 3 XP_005269439.1

View Full Product Details