Product Details

SNP ID
rs1042796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63488364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCCGGCGCCGCCGCTCTTCTT[C/T]TCCACGTTCTTGATGACGCCTACGG
Phenotype
MIM: 602959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1A2 PubMed Links

Gene Details

Gene
EEF1A2
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001958.3 1655 Silent Mutation GAA,GAG E442E NP_001949.1

View Full Product Details