Product Details

SNP ID
rs1467227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45892970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCAGCTTCGTTCTCAGCACTTA[A/G]AGGGCTGCAAAGAGGCAGGCTGGAT
Phenotype
MIM: 603732 MIM: 604641
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRY2 PubMed Links

Gene Details

Gene
CRY2
Gene Name
cryptochrome circadian clock 2
There are no transcripts associated with this gene.

Gene
MAPK8IP1
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005456.3 Intron NP_005447.1
XM_005253226.3 Intron XP_005253283.1

View Full Product Details