Product Details

SNP ID
rs1046990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86530758 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGACGCTGAGCTGAATGCCCA[A/G]TGACAAACGTGACCAGCCGTGCGGG
Phenotype
MIM: 616820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTHFSD PubMed Links

Gene Details

Gene
MTHFSD
Gene Name
methenyltetrahydrofolate synthetase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159377.1 2456 UTR 3 NP_001152849.1
NM_001159378.1 2456 UTR 3 NP_001152850.1
NM_001159379.1 2456 UTR 3 NP_001152851.1
NM_001159380.1 2456 UTR 3 NP_001152852.1
NM_022764.2 2456 UTR 3 NP_073601.2
XM_005256101.1 2456 UTR 3 XP_005256158.1
XM_005256105.1 2456 UTR 3 XP_005256162.1
XM_005256106.1 2456 UTR 3 XP_005256163.1
XM_006721247.3 2456 UTR 3 XP_006721310.1
XM_011523280.2 2456 UTR 3 XP_011521582.1
XM_011523282.2 2456 Intron XP_011521584.1
XM_011523283.2 2456 Intron XP_011521585.1
XM_011523284.2 2456 Intron XP_011521586.1
XM_011523285.1 2456 UTR 3 XP_011521587.1
XM_011523286.1 2456 UTR 3 XP_011521588.1
XM_011523287.1 2456 UTR 3 XP_011521589.1
XM_011523288.1 2456 UTR 3 XP_011521590.1
XM_011523289.1 2456 UTR 3 XP_011521591.1
XM_017023571.1 2456 UTR 3 XP_016879060.1
XM_017023572.1 2456 UTR 3 XP_016879061.1
XM_017023573.1 2456 UTR 3 XP_016879062.1
XM_017023574.1 2456 UTR 3 XP_016879063.1

View Full Product Details