Product Details

SNP ID
rs1804618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103337141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTCAATCCAAAGAAGCAAACAA[A/T]AGGTAATTCTTGTAAAGCCTGTGGC
Phenotype
MIM: 601710
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF5 PubMed Links

Gene Details

Gene
EIF5
Gene Name
eukaryotic translation initiation factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001969.4 886 Missense Mutation AAA,ATA K118I NP_001960.2
NM_183004.4 886 Missense Mutation AAA,ATA K118I NP_892116.2
Gene
LOC105370687
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SNORA28
Gene Name
small nucleolar RNA, H/ACA box 28
There are no transcripts associated with this gene.

View Full Product Details